Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Muc18B
Entrez Gene ID
32913
Gene Name
Mucin 18B
Gene Aliases
CG7876, Dmel\CG7876, Dmel_CG7876
Chromosome Location
Chr.X: 19215515 - 19216660 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031000
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298502.1 NP_001285431.1 1 - 1 1029 64
NM_133137.3 NP_573365.1 1 - 1 1029 64
GenBank mRNA AY075488.1 - 1 - 1 1027 64

View Full Product Details