Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mthl3
Entrez Gene ID
36961
Gene Name
methuselah-like 3
Gene Aliases
104/8, BEST:GM02553, CG6530, Dmel\CG6530, Dmel_CG6530, GENE 1, Mth-3, Mthl3, anon-WO0170980.52, anon-WO0170980.53, gene 1, l(2)k10408, mthl-3
Chromosome Location
Chr.2R: 17448533 - 17452598 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028956
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299597.1 NP_001286526.1 7 - 8 1914 60
NM_001299598.1 NP_001286527.1 7 - 8 1665 60
NM_145335.3 NP_663308.1 7 - 8 1603 60

View Full Product Details