Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ank
Entrez Gene ID
43770
Gene Name
Ankyrin
Gene Aliases
ANK, CG1651, CG32008, DAnk1, Dank1, Dmel\CG1651, Dmel_CG1651, ank, dank1
Chromosome Location
Chr.4: 115074 - 129849 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011747
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169348.2 NP_001162819.1 9 - 10 3085 87
NM_175926.3 NP_787122.1 8 - 9 3043 87
NM_175927.3 NP_787123.1 9 - 10 3118 87
NM_175928.3 NP_787124.1 9 - 10 3233 87
GenBank mRNA BT031123.1 - 8 - 9 3038 87

View Full Product Details