Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gr23a
- Entrez Gene ID
-
33453
- Gene Name
- Gustatory receptor 23a
- Gene Aliases
-
23A.1a, 23A1.b, 23A1a, 23A1b, CG15396, Dmel\CG15396, Dmel_CG15396, GR23A.1, GR23a, Gr23A1a, Gr23A1b, Gr23a.a, Gr23a.b, Gr23aA, Gr23aB
- Chromosome Location
-
Chr.2L: 2669823 - 2672186 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041248
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_175951.2 |
NP_787965.1 |
1 - 2 |
881 |
137 |
View Full Product Details