Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hoe1
Entrez Gene ID
249663
Gene Name
hoepel1
Gene Aliases
BcDNA:GH02636, CG11918, CG12787, CG2831, Dmel\CG12787, Dmel_CG12787, Hoe1, hoe, hoe-1
Chromosome Location
Chr.2L: 4908581 - 4932384 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041150
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273130.1 NP_001260059.1 2 - 3 714 57
NM_001273131.1 NP_001260060.1 2 - 3 672 57
NM_164603.2 NP_723024.2 2 - 3 600 57
NM_175966.2 NP_787980.1 2 - 3 684 57
NM_175967.2 NP_787981.1 2 - 3 722 57
NM_175968.2 NP_787982.1 2 - 3 668 57
GenBank mRNA AY050571.1 - 2 - 3 687 57
BT016131.1 - 2 - 3 687 57

View Full Product Details