Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slv
Entrez Gene ID
35773
Gene Name
saliva
Gene Aliases
CG8717, Dmel\CG8717, Dmel_CG8717
Chromosome Location
Chr.2R: 8080559 - 8083357 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025469
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273868.2 NP_001260797.1 4 - 5 802 75
NM_001299253.1 NP_001286182.1 4 - 5 668 75
NM_058035.4 NP_477383.1 3 - 4 512 75
GenBank mRNA AY118993.1 - 3 - 4 393 75

View Full Product Details