Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rep
Entrez Gene ID
37246
Gene Name
Rab escort protein
Gene Aliases
CG8432, Dmel\CG8432, Dmel_CG8432, REP
Chromosome Location
Chr.2R: 19492087 - 19494125 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026378
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058072.4 NP_477420.1 1 - 2 185 65
GenBank mRNA AY061052.1 - 1 - 2 127 65

View Full Product Details