Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stam
Entrez Gene ID
34505
Gene Name
Signal transducing adaptor molecule
Gene Aliases
CG6521, Dmel\CG6521, Dmel_CG6521, Dstam, STAM, Stam1, dstam, stam
Chromosome Location
Chr.2L: 10858536 - 10862006 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027363
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058100.4 NP_477448.1 3 - 4 1829 68
GenBank mRNA AF121359.1 - 3 - 4 1822 68
BT006014.1 - 3 - 4 1829 68

View Full Product Details