Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sur
Entrez Gene ID
34350
Gene Name
Sulfonylurea receptor
Gene Aliases
BEST:CK00325, CG5772, CK00325, CYO, DSur, Dme_CG5772, Dmel\CG5772, Dmel_CG5772, Dsur, Ir, SUR, cyo, dSUR, dSur, dsur, sur
Chromosome Location
Chr.2L: 10182557 - 10199117 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028675
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001347812.1 NP_001334732.1 21 - 22 6823 60

View Full Product Details