Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Fcp3C
- Entrez Gene ID
-
31294
- Gene Name
- Follicle cell protein 3C
- Gene Aliases
-
CG4015, Dmel\CG4015, Dmel_CG4015, EG:140G11.2, Ecp3C, VMP3C
- Chromosome Location
-
Chr.X: 3173376 - 3174474 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000644
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058134.5 |
NP_477482.2 |
2 - 3 |
739 |
61 |
GenBank mRNA |
BT088783.1 |
- |
3 - 4 |
1770 |
61 |
View Full Product Details