Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SkpD
- Entrez Gene ID
-
32995
- Gene Name
- SKP1-related D
- Gene Aliases
-
BcDNA:AT18217, CG12700, Dmel\CG12700, Dmel_CG12700, Skp, dskpD, skpD
- Chromosome Location
-
Chr.X: 19812729 - 19813384 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026174
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_134513.3 |
NP_608357.2 |
1 - 1 |
581 |
66 |
View Full Product Details