Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dop2R
Entrez Gene ID
33007
Gene Name
Dopamine 2-like receptor
Gene Aliases
CG17004, CG33517, CG9569, D2R, DD2R, DmDOP3, Dmel\CG33517, Dmel_CG33517, anon-WO0170980.130, anon-WO0170980.131, anon-WO0170980.31, anon-WO0170980.32, d2r, dD2R
Chromosome Location
Chr.X: 20004680 - 20062706 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0053517
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014757.3 NP_001014757.2 6 - 7 2510 107
NM_001014758.3 NP_001014758.2 6 - 7 2510 107
NM_001014759.2 NP_001014759.1 4 - 5 997 107
NM_001014760.3 NP_001014760.2 6 - 7 2510 107
NM_001031909.2 NP_001027080.2 6 - 7 2510 107
NM_001298548.1 NP_001285477.1 5 - 6 2394 107

View Full Product Details