Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AIF
Entrez Gene ID
33390
Gene Name
Apoptosis inducing factor
Gene Aliases
CG7263, DmAIF, Dmel\CG7263, Dmel_CG7263, SD03428p, aif
Chromosome Location
Chr.2L: 2151754 - 2155389 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031392
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272978.1 NP_001259907.1 1 - 2 395 109
NM_164456.3 NP_722765.2 1 - 2 395 109

View Full Product Details