Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Hrs
- Entrez Gene ID
-
33458
- Gene Name
- Hepatocyte growth factor regulated tyrosine kinase substrate
- Gene Aliases
-
CG2903, DmHRS, Dmel\CG2903, Dmel_CG2903, HRS, HRS-2, Vps27, dHrs, hrs, l(2)23AB5, l(2)23Ad, l(2)23Ad(Hrs), vps27
- Chromosome Location
-
Chr.2L: 2739986 - 2743377 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031450
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_164497.1 |
NP_722831.1 |
1 - 2 |
57 |
63 |
GenBank mRNA |
BT099905.1 |
- |
1 - 2 |
57 |
63 |
View Full Product Details