Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gyc32E
Entrez Gene ID
34553
Gene Name
Guanylyl cyclase at 32E
Gene Aliases
CG33114, CG6275, Cyg1, DMRGC, Dmel\CG33114, Dmel_CG33114, GC, GYC 32E, Gyc-r
Chromosome Location
Chr.2L: 11138759 - 11149101 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010197
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103678.2 NP_001097148.1 15 - 16 2054 119
NM_001298905.1 NP_001285834.1 14 - 15 1946 119
NM_176013.2 NP_788027.1 14 - 15 1970 119

View Full Product Details