Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sns
Entrez Gene ID
44097
Gene Name
sticks and stones
Gene Aliases
43-49, CG12495, CG13752, CG13753, CG13754, CG13755, CG18464, CG2385, CG33141, CG8278, CT33230, CT33232, Dmel\CG33141, Dmel_CG33141, SNS, Sns, rost, sns20
Chromosome Location
Chr.2R: 8798489 - 8856091 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024189
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043067.2 NP_001036532.1 21 - 22 4545 85
NM_001299284.1 NP_001286213.1 20 - 21 4395 85
NM_176106.2 NP_788286.1 20 - 21 4356 85

View Full Product Details