The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144160.2 | NP_001137632.1 | 14 - 15 | 2371 | 103 |
NM_001273919.1 | NP_001260848.1 | 14 - 15 | 2101 | 103 | |
NM_001273920.1 | NP_001260849.1 | 15 - 16 | 2155 | 103 | |
NM_001273921.1 | NP_001260850.1 | 15 - 16 | 2203 | 103 | |
NM_001273922.1 | NP_001260851.1 | 14 - 15 | 2167 | 103 | |
NM_176119.5 | NP_788299.3 | 14 - 15 | 2110 | 103 | |
NM_176120.5 | NP_788300.3 | 16 - 17 | 2665 | 103 | |
GenBank mRNA | AY823300.1 | - | 14 - 15 | 2104 | 103 |
AY823301.1 | - | 14 - 15 | 2104 | 103 |