Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
grau
Entrez Gene ID
45871
Gene Name
grauzone
Gene Aliases
CG3282, CG33133, Dmel\CG33133, Dmel_CG33133, Grau
Chromosome Location
Chr.2R: 21009764 - 21011921 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001133
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176242.3 NP_788422.1 1 - 2 201 71
GenBank mRNA AF208016.1 - 1 - 2 183 71
AY047537.1 - 1 - 2 168 71

View Full Product Details