Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gmap
Entrez Gene ID
32483
Gene Name
Golgi microtubule-associated protein
Gene Aliases
CG32587, CG33206, CG6350, CG7821, Dmel\CG33206, Dmel_CG33206, GMAP, GMAP-210, GMAP210, dGMAP, l(1)G0168
Chromosome Location
Chr.X: 15486922 - 15511223 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027287
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298345.1 NP_001285274.1 5 - 6 3835 79
NM_001370008.1 NP_001356946.1 6 - 7 4167 79
NM_176734.2 NP_788907.1 6 - 7 4167 79
NM_176735.3 NP_788908.1 5 - 6 3975 79
GenBank mRNA AY052121.1 - 6 - 7 4167 79

View Full Product Details