Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ranbp16
Entrez Gene ID
118436
Gene Name
CG33180-PB
Gene Aliases
CG33180, CG9126, CG9136, Dmel\CG33180, Dmel_CG33180, RanBP16
Chromosome Location
Chr.X: 15927185 - 15933867 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0053180
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201665.2 NP_001188594.1 11 - 12 2911 118
NM_001201666.2 NP_001188595.1 11 - 12 2911 118
NM_001201667.2 NP_001188596.1 11 - 12 2911 118
NM_001201668.2 NP_001188597.1 10 - 11 2973 118
NM_001298379.1 NP_001285308.1 11 - 12 3103 118
NM_176740.4 NP_788913.2 11 - 12 2911 118
GenBank mRNA AF222744.1 - 11 - 12 2893 118
BT021400.1 - 11 - 12 2886 118

View Full Product Details