Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sas
Entrez Gene ID
40861
Gene Name
stranded at second
Gene Aliases
CG2507, Dmel\CG2507, Dmel_CG2507, SAS, Sas, l(3)84Cd, rfd
Chromosome Location
Chr.3R: 7162808 - 7183702 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0002306
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275407.2 NP_001262336.1 2 - 3 458 58
NM_001275408.2 NP_001262337.1 3 - 4 896 58
NM_057263.5 NP_476611.1 3 - 4 810 58
NM_169177.3 NP_731141.1 2 - 3 576 58
GenBank mRNA AY051979.1 - 2 - 3 637 58

View Full Product Details