Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
osk
Entrez Gene ID
41066
Gene Name
oskar
Gene Aliases
CG10901, Dm-osk, Dmel\CG10901, Dmel_CG10901, OSK, Osk, Oskar, norka
Chromosome Location
Chr.3R: 8935126 - 8938395 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003015
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_169248.4 NP_731295.1 1 - 2 555 66
NM_206464.3 NP_996186.1 1 - 2 555 66
GenBank mRNA AY058560.1 - 1 - 2 531 66

View Full Product Details