Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- rod
- Entrez Gene ID
-
43719
- Gene Name
- rough deal
- Gene Aliases
-
CG1569, Dmel\CG1569, Dmel_CG1569, EP3408, ROD, Rod, Rod1, pNB20A
- Chromosome Location
-
Chr.3R: 31580951 - 31588244 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003268
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057357.3 |
NP_476705.1 |
5 - 6 |
1261 |
87 |
View Full Product Details