Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
trp
Entrez Gene ID
43542
Gene Name
transient receptor potential
Gene Aliases
559, CG7875, DmTRP, Dmel\CG7875, Dmel_CG7875, TRP, Trp, dTRP, dTRPC1, dTrp, gene B, idl-B
Chromosome Location
Chr.3R: 29914521 - 29920289 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003861
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276161.1 NP_001263090.1 5 - 6 1161 72
NM_057420.4 NP_476768.1 5 - 6 1141 72
GenBank mRNA BT150384.1 - 5 - 6 1143 72

View Full Product Details