Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptp99A
Entrez Gene ID
43469
Gene Name
Protein tyrosine phosphatase 99A
Gene Aliases
CG11515, CG11516, CG11517, CG2005, CT6383, DPTP99A, DPTP[[99A]], Dmel\CG11516, Dmel_CG11516, PTP99A, R-PTP 99A, dptp99A, ptp99A
Chromosome Location
Chr.3R: 29377646 - 29487131 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004369
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057468.3 NP_476816.1 3 - 4 1190 76
NM_143434.5 NP_651691.4 10 - 11 2207 76
NM_170409.3 NP_733288.1 4 - 5 1321 76
NM_206579.3 NP_996302.1 4 - 5 1366 76

View Full Product Details