Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zfh1
Entrez Gene ID
43650
Gene Name
Zn finger homeodomain 1
Gene Aliases
CG1322, Dmel\CG1322, Dmel_CG1322, ZFH-1, ZFH1, ZFh1, Zfh-1, Zfh1, Zfh1a, l(3)00865, zfh-1, zfl-1, zhf1
Chromosome Location
Chr.3R: 30765926 - 30789202 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004606
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057502.5 NP_476850.1 1 - 2 453 89
NM_170522.4 NP_733401.3 1 - 2 453 89

View Full Product Details