Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gl
Entrez Gene ID
42210
Gene Name
glass
Gene Aliases
CG7672, Dmel\CG7672, Dmel_CG7672, E(sina)5, Gl, Gla, Glass, SS3-2, SY3-3, gla, none, rh
Chromosome Location
Chr.3R: 18367927 - 18372738 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004618
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275779.1 NP_001262708.1 3 - 4 491 129
NM_057506.3 NP_476854.1 3 - 4 491 129
NM_169807.4 NP_732330.1 3 - 4 491 129
GenBank mRNA AY060322.1 - 3 - 4 470 129

View Full Product Details