Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- C15
- Entrez Gene ID
-
42544
- Gene Name
- C15-PA
- Gene Aliases
-
311, 93Bal, CG7937, Cll, Dmel\CG7937, Dmel_CG7937, Ect5, Hox11-311, c15, cll
- Chromosome Location
-
Chr.3R: 21499986 - 21512814 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004863
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057525.3 |
NP_476873.2 |
2 - 3 |
863 |
79 |
GenBank mRNA |
BT025203.2 |
- |
2 - 3 |
863 |
79 |
View Full Product Details