Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ChAT
- Entrez Gene ID
-
42249
- Gene Name
- Choline acetyltransferase
- Gene Aliases
-
CG12345, CHAT, CLAT_DROME, CT23399, Cat, ChA, ChAt, Cha, ChaT, DChAT, Dmel\CG12345, Dmel_CG12345, cha, chat, dChAT, l(3)91Cc
- Chromosome Location
-
Chr.3R: 18705501 - 18732344 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000303
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057656.3 |
NP_477004.5 |
6 - 7 |
2193 |
66 |
GenBank mRNA |
M63724.2 |
- |
6 - 7 |
2171 |
66 |
View Full Product Details