Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pug
Entrez Gene ID
41279
Gene Name
pugilist
Gene Aliases
CG4067, Dmel\CG4067, Dmel_CG4067, MTHFD, Pug, bs14h02.y1, pugD
Chromosome Location
Chr.3R: 10696534 - 10703316 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020385
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014614.2 NP_001014614.1 2 - 3 227 86
NM_001260099.1 NP_001247028.1 2 - 3 185 86
NM_057906.4 NP_477254.1 1 - 2 165 86
NM_169350.3 NP_731489.2 2 - 3 340 86
NM_169351.2 NP_731490.1 2 - 3 111 86
GenBank mRNA BT125950.1 - 2 - 3 210 86

View Full Product Details