Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pug
Entrez Gene ID
41279
Gene Name
pugilist
Gene Aliases
CG4067, Dmel\CG4067, Dmel_CG4067, MTHFD, Pug, bs14h02.y1, pugD
Chromosome Location
Chr.3R: 10696534 - 10703316 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020385
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014614.2 NP_001014614.1 5 - 6 2747 72
NM_001260099.1 NP_001247028.1 5 - 6 2705 72
NM_057906.4 NP_477254.1 4 - 5 2685 72
NM_169350.3 NP_731489.2 5 - 6 2860 72
NM_169351.2 NP_731490.1 5 - 6 2631 72
GenBank mRNA BT125950.1 - 5 - 6 2730 72

View Full Product Details