Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mira
- Entrez Gene ID
-
42379
- Gene Name
- miranda
- Gene Aliases
-
CG12249, Dmel\CG12249, Dmel_CG12249, MIR, MIRA, Mir, Mira, Miranda, mir
- Chromosome Location
-
Chr.3R: 19931799 - 19935120 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0021776
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057943.4 |
NP_477291.1 |
1 - 1 |
2148 |
98 |
View Full Product Details