Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
amon
Entrez Gene ID
43215
Gene Name
amontillado
Gene Aliases
Amon, CG6438, DMH#5, Dmel\CG6438, Dmel_CG6438, PC2, PPP, Ppp, dPC2
Chromosome Location
Chr.3R: 26697697 - 26714159 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0023179
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057970.4 NP_477318.1 11 - 12 2242 96
GenBank mRNA AY119509.1 - 11 - 12 2200 96

View Full Product Details