Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Lip3
- Entrez Gene ID
-
41643
- Gene Name
- CG8823-PA
- Gene Aliases
-
CG8823, Dilp3, Dmel\CG8823, Dmel_CG8823, Dmlip3, LIP3, LIP3Dm, dLip3, dilp3, lip3
- Chromosome Location
-
Chr.3R: 13370240 - 13371904 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0023495
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057983.4 |
NP_477331.1 |
1 - 2 |
236 |
65 |
GenBank mRNA |
BT023257.1 |
- |
1 - 2 |
236 |
65 |
View Full Product Details