Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Irp-1A
- Entrez Gene ID
-
42689
- Gene Name
- Iron regulatory protein 1A
- Gene Aliases
-
C-Acon, CG4900, Dmel\CG4900, Dmel_CG4900, IRF, IRP, IRP-1, IRP-1A, IRP1, IRP1A, c-Acon, cACON, cAcon, dIRP, irp1A
- Chromosome Location
-
Chr.3R: 22721320 - 22724587 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0024958
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058023.5 |
NP_477371.1 |
2 - 3 |
175 |
56 |
GenBank mRNA |
AY058657.1 |
- |
2 - 3 |
124 |
56 |
View Full Product Details