Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Manf
- Entrez Gene ID
-
41983
- Gene Name
- Mesencephalic astrocyte-derived neurotrophic factor
- Gene Aliases
-
ARP-like, CG7013, DMANF, DmMANF, DmManf, Dmel\CG7013, Dmel_CG7013, MANF, anon-EST:ParkEST188, manf
- Chromosome Location
-
Chr.3R: 16349404 - 16350801 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0027095
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058097.6 |
NP_477445.1 |
2 - 3 |
325 |
97 |
GenBank mRNA |
AY061080.1 |
- |
2 - 3 |
312 |
97 |
View Full Product Details