Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hkb
Entrez Gene ID
40549
Gene Name
huckebein
Gene Aliases
5953, CG9768, Dmel\CG9768, Dmel_CG9768, HKB, Hkb, anon-82B3, gurt
Chromosome Location
Chr.3R: 4345015 - 4346646 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261434
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079497.4 NP_524221.1 1 - 2 801 63
GenBank mRNA AY113527.1 - 1 - 2 797 63

View Full Product Details