Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
alpha-Est8
Entrez Gene ID
40899
Gene Name
alpha-Esterase-8
Gene Aliases
CG1121, DmalphaE8, Dmel\CG1121, Dmel_CG1121, Est8, I, aE8, alphaE8
Chromosome Location
Chr.3R: 7509343 - 7515670 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015576
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079535.3 NP_524259.2 3 - 4 1707 126
GenBank mRNA BT150138.1 - 3 - 4 1706 126

View Full Product Details