Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or85c
- Entrez Gene ID
-
41008
- Gene Name
- Odorant receptor 85c
- Gene Aliases
-
85A.3, 85c, CG17911, DOR78, Dmel\CG17911, Dmel_CG17911
- Chromosome Location
-
Chr.3R: 8511734 - 8513022 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037591
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079556.3 |
NP_524280.2 |
1 - 2 |
959 |
93 |
View Full Product Details