Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FER
Entrez Gene ID
41118
Gene Name
FER tyrosine kinase
Gene Aliases
CG8874, DFER, DFer, Dfer, Dfps, Dm FPS, DmHD-179, Dmel\CG8874, Dmel_CG8874, FPS, Fer, FpS95D, Fps85D, HD-179, dFer, dfer, dfps, dfps 85D, dfps85D, fer, fps, fps85D
Chromosome Location
Chr.3R: 9367603 - 9394546 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000723
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275482.1 NP_001262411.1 2 - 3 537 72
NM_001275483.1 NP_001262412.1 2 - 3 537 72
NM_079564.4 NP_524288.3 2 - 3 537 72
NM_169274.2 NP_731341.1 2 - 3 537 72
GenBank mRNA BT003462.1 - 2 - 3 535 72

View Full Product Details