Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- hyd
- Entrez Gene ID
-
41181
- Gene Name
- hyperplastic discs
- Gene Aliases
-
CG9484, Dmel\CG9484, Dmel_CG9484, HYD, Hyd, NP_524296.2, UBE 3A, UBR5, c43, l(3)85Ea, l(3)C43, l(3)C43-3, l(3)SG33, l(3)c43, l[[3]]c43
- Chromosome Location
-
Chr.3R: 9714623 - 9725615 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0002431
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079572.4 |
NP_524296.2 |
7 - 8 |
4776 |
66 |
GenBank mRNA |
BT150484.1 |
- |
7 - 8 |
4760 |
66 |
View Full Product Details