Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mfas
Entrez Gene ID
41455
Gene Name
midline fasciclin
Gene Aliases
87A7-9/7, BEST:GH06752, CG3359, Dmel\CG3359, Dmel_CG3359, Mfas, anon-87Af, anon-EST:Liang-97, anon-WO0140519.174, clone 97, gh06752
Chromosome Location
Chr.3R: 11984862 - 11993315 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260745
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260137.1 NP_001247066.1 5 - 6 895 59
NM_079600.3 NP_524324.2 6 - 7 1225 59
NM_169444.1 NP_731661.1 6 - 7 913 59
NM_176466.1 NP_788643.1 6 - 7 932 59
NM_176467.1 NP_788644.1 6 - 7 910 59
NM_176468.1 NP_788645.1 5 - 6 1309 59
NM_176469.2 NP_788646.1 6 - 7 1030 59
NM_176470.1 NP_788647.1 6 - 7 935 59
NM_176471.1 NP_788648.1 6 - 7 1108 59
NM_176472.1 NP_788649.1 6 - 7 1130 59
NM_176473.1 NP_788650.1 7 - 8 1004 59
NM_176474.1 NP_788651.1 7 - 8 982 59
NM_176475.1 NP_788652.1 6 - 7 1381 59
NM_176476.1 NP_788653.1 7 - 8 985 59
NM_176477.1 NP_788654.1 7 - 8 1007 59
NM_176478.1 NP_788655.1 7 - 8 1180 59
GenBank mRNA AY069094.1 - 6 - 7 895 59
BT120190.1 - 5 - 6 895 59
BT125878.1 - 7 - 8 981 59
BT133296.1 - 7 - 8 1159 59

View Full Product Details