Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Hug
- Entrez Gene ID
-
41547
- Gene Name
- Hugin
- Gene Aliases
-
CG6371, Dmel\CG6371, Dmel_CG6371, Drm-MT2, Drm-PK-2, HUG, HUGgamma, Hug-PK, Hug-gamma, Hugin-PK-2, PK-2, hug, hug gamma, hug-1, hug-2, hug-PK, hug-gamma, hugg, huggamma, hugin, huginPK, pyrokinin/PBAN-like
- Chromosome Location
-
Chr.3R: 12526188 - 12529076 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0028374
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079605.2 |
NP_524329.1 |
2 - 3 |
535 |
70 |
GenBank mRNA |
AY047528.1 |
- |
2 - 3 |
515 |
70 |
View Full Product Details