Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mbo
- Entrez Gene ID
-
41562
- Gene Name
- members only
- Gene Aliases
-
1465/07, 1465/7, CG6819, DNup88, Dmel\CG6819, Dmel_CG6819, Dnup 88, Dnup88, MBO, Mbo, NUP88, Nup88, anon-WO0118547.263, dNup88, l(3)05043, l(3)S146507, nup88
- Chromosome Location
-
Chr.3R: 12676487 - 12679052 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026207
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079606.4 |
NP_524330.2 |
1 - 1 |
2331 |
99 |
GenBank mRNA |
AY051827.1 |
- |
1 - 1 |
2310 |
99 |
View Full Product Details