Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- timeout
- Entrez Gene ID
-
41615
- Gene Name
- CG7855-PA
- Gene Aliases
-
CG14381, CG14382, CG7855, CG8148, Dmel\CG7855, Dmel_CG7855, TIM2, Tim-2, dtim2, tim, tim2
- Chromosome Location
-
Chr.3R: 13088651 - 13163876 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038118
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079617.3 |
NP_524341.3 |
15 - 16 |
2576 |
90 |
View Full Product Details