Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tm1
Entrez Gene ID
41852
Gene Name
Tropomyosin 1
Gene Aliases
1305/10, 2299, BcDNA:GH09289, BcDNA:LD37158, BcDNA:SD21996, CG4898, Dm Tm1, Dm TmH33, Dm TmH34, DmTm1, Dmel\CG4898, Dmel_CG4898, PmI, TM, TM1, TMII, Tm, TmH, TmH-33, TmH-34, TmH33, TmH34, TmII, Tmr33, Tmr34, TnH, TnH-33, TnH-34, cTM, cTm, cTmII, chr3R:11122272..11122408, l(3)02299, l(3)S130510, l(3)s2958, mTmII, region 3, tm1, tmII, tropomyosin
Chromosome Location
Chr.3R: 15278220 - 15308779 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003721
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382175.1 NP_001369014.1 7 - 8 1017 60
NM_079636.4 NP_524360.2 5 - 6 982 60
NM_206494.3 NP_996216.1 7 - 8 730 60
GenBank mRNA AY060414.1 - 5 - 6 962 60

View Full Product Details