Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sap47
Entrez Gene ID
41938
Gene Name
Synapse-associated protein 47kD
Gene Aliases
CG8884, Dmel\CG8884, Dmel_CG8884, SAP 47, SAP-47, SAP47, dsap-47, sap47
Chromosome Location
Chr.3R: 15935100 - 15962124 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013334
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300421.1 NP_001287350.1 2 - 3 1489 71
NM_001370053.1 NP_001356917.1 2 - 3 1489 71
NM_001370054.1 NP_001356918.1 3 - 4 1102 71
NM_079647.4 NP_524371.1 2 - 3 1470 71
NM_169684.3 NP_732088.1 3 - 4 1518 71
NM_169685.3 NP_732089.1 4 - 5 1126 71
NM_169686.3 NP_732090.1 4 - 5 1454 71
NM_169687.3 NP_732091.1 4 - 5 1272 71
NM_169688.3 NP_732092.2 2 - 3 1489 71
NM_169689.3 NP_732093.1 3 - 4 1102 71
NM_169690.3 NP_732094.1 3 - 4 1518 71
GenBank mRNA AY051871.1 - 3 - 4 1466 71
X80110.1 - - 761 71
X80111.1 - - 1089 71

View Full Product Details