Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rbf2
Entrez Gene ID
41941
Gene Name
Retinoblastoma-family protein 2
Gene Aliases
CG5083, Dmel\CG5083, Dmel_CG5083, RB, RBF2, RBf2, rbf2
Chromosome Location
Chr.3R: 15965446 - 15968138 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038390
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079648.4 NP_524372.1 1 - 1 2341 65
GenBank mRNA AF197059.1 - 1 - 1 2278 65
AY071301.1 - 1 - 1 2336 65

View Full Product Details