Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cher
Entrez Gene ID
42066
Gene Name
cheerio
Gene Aliases
CG3937, Cher, Dmel\CG3937, Dmel_CG3937, FLN, FLN90, Fil, filamin, joy, sko
Chromosome Location
Chr.3R: 17091533 - 17125389 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0014141
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202309.1 NP_001189238.1 12 - 13 4462 92
NM_001275728.1 NP_001262657.2 14 - 15 5339 92
NM_001275729.1 NP_001262658.1 13 - 14 5029 92
NM_001275730.1 NP_001262659.2 15 - 16 5354 92
NM_079659.4 NP_524383.3 12 - 13 4523 92
NM_206502.3 NP_996224.2 13 - 14 5090 92
GenBank mRNA AF188360.1 - 12 - 13 4499 92

View Full Product Details