Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
beat-IIa
Entrez Gene ID
42086
Gene Name
beaten path IIa
Gene Aliases
BcDNA:RE17794, Beat-IIa, CG14334, CG14335, CT33965, Dmel\CG14334, Dmel_CG14334, beat IIa
Chromosome Location
Chr.3R: 17320004 - 17366119 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038498
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300446.1 NP_001287375.1 1 - 2 938 84
NM_079660.4 NP_524384.3 2 - 3 1093 84
GenBank mRNA AF325713.1 - 2 - 3 892 84
AY118373.1 - 2 - 3 1091 84
BT023786.1 - 1 - 2 189 84

View Full Product Details